Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 3A7 Antibody (F19 P2 H2), Novus Biologicals™
SDP

Catalog No. NB10074402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074402 Supplier Novus Biologicals Supplier No. NB10074402

Mouse Monoclonal Antibody

Cytochrome P450 3A7 Monoclonal specifically detects Cytochrome P450 3A7 in Human, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytochrome P450 3A7
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone F19 P2 H2
Conjugate Unconjugated
Dilution Western Blot 1:250, Flow Cytometry 2 ug 10^6 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500
Formulation Ascites with 0.05% Sodium Azide
Gene Alias Aryl hydrocarbon hydroxylase, CP37, CYPIIIA7, cytochrome P450, Cytochrome P450 3A7, family 3, Flavoprotein linked monooxygenase, Microsomal monooxygenase, P450 HFLA, polypeptide 7, subfamily A, subfamily IIIA, Xenobiotic monooxygenase
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues E S R D E T V S G A, conjugated to ovalbumin.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Cytochrome P450 3A7 (F19 P2 H2)
Target Species Human, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.