Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 4A Antibody (M25-P2A10), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP250212AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250212AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250212AG Supplier Novus Biologicals Supplier No. NBP250212AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 4A Monoclonal specifically detects Cytochrome P450 4A in Human, Mouse, Rat, Drosophila, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Cytochrome P450 4A
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M25-P2A10
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 20-HETE synthase, CP4Y, CYP4A2, cytochrome P450 4A11, cytochrome P450, family 4, subfamily A, polypeptide 11, cytochrome P450, subfamily IVA, polypeptide 11, cytochrome P-450HK-omega, cytochrome P450HL-omega, fatty acid omega-hydroxylase, lauric acid omega-hydroxylase, P450HL-omega
Gene Symbols CYP4A11
Host Species Mouse
Immunogen Peptide Sequence - KNGIHLRLR (amino acids 499 - 507)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1579
Target Species Human, Mouse, Rat, Drosophila, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.