Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 4A1, 2C11 Antibody (Clo1), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP250211C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250211C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250211C Supplier Novus Biologicals Supplier No. NBP250211C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 4A1, 2C11 Monoclonal specifically detects Cytochrome P450 4A1, 2C11 in Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Cytochrome P450 4A1, 2C11
Applications Western Blot, ELISA
Classification Monoclonal
Clone Clo1
Conjugate DyLight 650
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CYP4A1
Host Species Mouse
Immunogen Rat liver cytochrome P450 2C11 and 4A1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.