Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 4A2, 4A3 Antibody (Clo4), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP20213A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20213A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20213A70 Supplier Novus Biologicals Supplier No. NBP250213AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 4A2, 4A3 Monoclonal specifically detects Cytochrome P450 4A2, 4A3 in Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Cytochrome P450 4A2, 4A3
Applications Western Blot, ELISA
Classification Monoclonal
Clone Clo4
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Rat liver cytochrome P450 4A2 and 4A3
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.