Learn More
Description
Specifications
Specifications
| Antigen | CytochromeP4504F11 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | F21 P6 F5 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 1-5 ug/ml, Immunohistochemistry-Paraffin 1-5 ug/ml |
| Formulation | PBS with 0.08% Sodium Azide |
| Gene Accession No. | A8K059 |
| Gene Alias | CYPIVF11, cytochrome P450 4F11, cytochrome P450, family 4, subfamily F, polypeptide 11, cytochrome P450, subfamily IVF, polypeptide 11, EC 1.14.14.1 |
| Gene Symbols | CYP4F11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
