Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 Enzyme CYP3A1 Antibody (P6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB390828 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390828 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390828 Supplier Novus Biologicals Supplier No. NBP308767AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 Enzyme CYP3A1 Monoclonal antibody specifically detects Cytochrome P450 Enzyme CYP3A1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen Cytochrome P450 Enzyme CYP3A1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone P6
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Full length protein (Rat). Rat liver cytochrome Cytochrome P450 Enzyme CYP3A1.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.