Learn More
Cytochrome P450 Enzyme CYP3A1 Antibody (P6) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytochrome P450 Enzyme CYP3A1 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | P6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 1-2 ug/mL, Flow Cytometry : 1-2 ug/million cells, ELISA |
| Formulation | 10mM PBS |
| Host Species | Mouse |
| Immunogen | Full length protein (Rat). Rat liver cytochrome Cytochrome P450 Enzyme CYP3A1. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.