Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytohesin-1 Antibody (2E11), Novus Biologicals™
SDP

Catalog No. NBP222512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222512 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222512 Supplier Novus Biologicals Supplier No. NBP222512

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Cytohesin-1 Monoclonal specifically detects Cytohesin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cytohesin-1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 200000000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Flow Cytometry 3 - 5 ug 10^6 cells, Immunocytochemistry/Immunofluorescence 1:20
Gene Accession No. Q15438
Gene Alias B2-1, cytoadhesin 1, cytohesin 1, cytohesin-1, D17S811ECYTOHESIN-1, FLJ34050, FLJ41900, homolog of secretory protein SEC7, PH, SEC7 and coiled-coil domain-containing protein 1, pleckstrin homology, Sec7 and coiled-coil domains 1, PSCD1, SEC7, SEC7 homolog B2-1
Gene Symbols CYTH1
Host Species Mouse
Immunogen Purified, His-tagged cytohesin-1.
Purification Method PEG purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 9267
Test Specificity This antibody reacts with Human.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.