Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytohesin 2 Antibody (CYT2-21) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB12012421 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12012421 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12012421 Supplier Novus Biologicals Supplier No. NB12012421
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytohesin 2 Monoclonal specifically detects Cytohesin 2 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cytohesin 2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CYT2-21
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Alias ARF exchange factor, ARF nucleotide-binding site opener, ARNOPH, SEC7 and coiled-coil domain-containing protein 2, CTS18.1, cytohesin 2, pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2), Protein ARNO, PSCD2LCTS18, PSCD2pleckstrin homology, Sec7 and coiled-coil domains 2, Sec7 and coiled/coil domains 2 (cytohesin-2)
Gene Symbols CYTH2
Host Species Mouse
Immunogen Recombinant full length protein (Human).
Purification Method Protein A or G purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 9266
Test Specificity does not cross-react with other members of the cytohesin family.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.