Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytohesin 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | Cytohesin 3 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cytohesin 3 Polyclonal specifically detects Cytohesin 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytohesin 3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
ARF nucleotide-binding site opener 3, ARNO3PH, SEC7 and coiled-coil domain-containing protein 3, cytohesin 3, General receptor of phosphoinositides 1, Grp1, GRP1cytohesin-3, pleckstrin homology, Sec7 and coiled-coil domains 3, Protein ARNO3, PSCD3pleckstrin homology, Sec7 and coiled/coil domains 3 | |
CYTH3 | |
IgG | |
Affinity Purified | |
Specificity of human Cytohesin 3 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9265 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DEDGGGEGGGVPEDLSLEEREELLDIRRRKKELIDDIERLKYEIAEVMTEIDNLTSVEESKTTQRNKQIAM | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title