Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 1 Antibody (LHK1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB1002756G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002756G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002756G Supplier Novus Biologicals Supplier No. NB1002756G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 1 Monoclonal specifically detects Cytokeratin 1 in Human, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Cytokeratin 1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LHK1
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. P04264
Gene Alias CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1
Gene Symbols KRT1
Host Species Mouse
Immunogen C-terminal peptide sequence of human Cytokeratin 1 [UniProt# P04264]
Molecular Weight of Antigen 67 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3848
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.