Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 10 Antibody (18B6), Novus Biologicals™
SDP

Catalog No. NB298621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298621 100 μL
NB298620 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298621 Supplier Novus Biologicals Supplier No. NBP326178100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen Cytokeratin 10
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 18B6
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis, keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Cytokeratin 10 [UniProt P13645]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3858
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.