Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 10 Antibody (18B6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Cytokeratin 10 |
---|---|
Clone | 18B6 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Cytokeratin 10 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis, keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
A synthesized peptide derived from Human Cytokeratin 10 [UniProt P13645] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
18B6 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3858 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title