Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 10 Antibody (KRT10/1948R), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB378981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378981 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378981 Supplier Novus Biologicals Supplier No. NBP308716C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human, Canine, Feline samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin 10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT10/1948R
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP
Host Species Rabbit
Immunogen Recombinant human full-length Cytokeratin 10 protein (Uniprot: P13645)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3858
Target Species Human, Canine, Feline
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.