Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 10 Antibody (KRT10/1990R), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB009030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009030 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009030 Supplier Novus Biologicals Supplier No. NBP275768AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 1/10
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT10/1990R
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 67 kDa cytokeratin, CK 1, CK 10, CK1, CK10, Cytokeratin 10, Cytokeratin1, Cytokeratin10, EHK 1, EHK1, EPPK, Hair Alpha Protein, K1, k10, Keratin 1, Keratin 1 (epidermolytic hyperkeratosis), Keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), Keratin type I cytoskeletal 10, Keratin Type II Cytoskeletal 1, Keratin1, Keratin10, KPP, KRT 1, Krt 10, KRT1, KRT10, KRT1A, KRTA, NEPPK
Gene Symbols KRT10
Host Species Rabbit
Immunogen Recombinant human full-length Cytokeratin 10 protein (Uniprot: P13645)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Test Specificity This MAb recognizes a protein of 56.5 kDa, identified as cytokeratin 10 (CK10). CK10 is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. However, CK10 is rarely detected in early stages of vulvar squamous carcinomas (tumors less than 2 cm, clinical stage I) regardless of the tumor grade. In larger and more advanced tumors (greater than 2 cm, clinical stages II and III), CK10 is detected very frequently. Expression of CK10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.