Learn More
Cytokeratin 10 Antibody (KRT10/1990R), DyLight 755, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytokeratin 1/10 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | KRT10/1990R |
| Conjugate | DyLight 755 |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | 67 kDa cytokeratin, CK 1, CK 10, CK1, CK10, Cytokeratin 10, Cytokeratin1, Cytokeratin10, EHK 1, EHK1, EPPK, Hair Alpha Protein, K1, k10, Keratin 1, Keratin 1 (epidermolytic hyperkeratosis), Keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), Keratin type I cytoskeletal 10, Keratin Type II Cytoskeletal 1, Keratin1, Keratin10, KPP, KRT 1, Krt 10, KRT1, KRT10, KRT1A, KRTA, NEPPK |
| Gene Symbols | KRT10 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.