Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 13 Antibody (KRT13/2850), Biotin, Novus Biologicals™
SDP

Catalog No. NB381976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381976 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381976 Supplier Novus Biologicals Supplier No. NBP308288B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 13 Monoclonal antibody specifically detects Cytokeratin 13 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin 13
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT13/2850
Conjugate Biotin
Formulation PBS
Gene Alias CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781
Host Species Mouse
Immunogen esophageal keratins of rabbit origin
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3860
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.