Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 14, Mouse anti-Human, Clone: A2C11, Novus Biologicals™
SDP

Catalog No. NB013476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013476 Supplier Novus Biologicals Supplier No. NBP280404

Mouse Monoclonal Antibody

Cytokeratin 14 Monoclonal specifically detects Cytokeratin 14 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 14
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A2C11
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200Flow 1:50-1:100
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ
Gene Symbols KRT14
Host Species Mouse
Immunogen Synthetic peptide within Human Cytokeratin 14 aa 423-472/ 472. (SwissProt: P02533 Human)
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3861
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.