Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 15 Antibody (KRT15/2103R), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB379546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379546 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379546 Supplier Novus Biologicals Supplier No. NBP308609AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytokeratin 15 Monoclonal antibody specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin 15
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT15/2103R
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15
Host Species Rabbit
Immunogen Recombinant human full-length Cytokeratin 15 protein (Uniprot: P19012)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3866
Target Species Human, Rat, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.