Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 16 Antibody (OTI2H8), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP271085UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271085UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271085UV Supplier Novus Biologicals Supplier No. NBP271085UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 16 Monoclonal specifically detects Cytokeratin 16 in Human, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen Cytokeratin 16
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI2H8
Conjugate DyLight 350
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK
Gene Symbols KRT16
Host Species Mouse
Immunogen Full length recombinant protein of human Cytokeratin 16 (NP_005548) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3868.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.