Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 16 Antibody (rKRT16/1714), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB392062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392062 Supplier Novus Biologicals Supplier No. NBP308539AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 16 Monoclonal antibody specifically detects Cytokeratin 16 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin 16
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rKRT16/1714
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK
Host Species Mouse
Immunogen Recombinant fragment from the C-terminal of human Cytokeratin 16 (Uniprot: P08779)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3868
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.