Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 17 Mouse anti-Human, Rat, Porcine, Bovine, Goat, Biotin, Clone: E3 (same as Ks17.E3), Novus Biologicals™
SDP

Catalog No. NB019608 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019608 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019608 Supplier Novus Biologicals Supplier No. NBP233188B100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 17 Monoclonal specifically detects Cytokeratin 17 in Human, Rat, Porcine, Bovine, Goat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Cytokeratin 17
Applications ELISA
Classification Monoclonal
Clone E3 (same as Ks17.E3)
Conjugate Biotin
Formulation PBS
Gene Alias 39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1
Host Species Mouse
Immunogen The cytoskeletal fraction of rat colon epithelium was used as immunogen for the Cytokeratin 17 E3 antibody.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3872
Target Species Human, Rat, Pig, Bovine, Goat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.