Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 18 Antibody (C-04), PerCP, Novus Biologicals™
SDP

Catalog No. NB500306PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500306PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500306PCP Supplier Novus Biologicals Supplier No. NB500306PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Mouse, Mammal samples. It is validated for Flow Cytometry.

Specifications

Antigen Cytokeratin 18
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone C-04
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18
Gene Symbols KRT18
Host Species Mouse
Immunogen Cytoskeleton preparation of epidermal carcinoma cell line A431.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3875
Test Specificity The antibody C-04 reacts with Cytokeratin 18 (45 kDa), a member of intermediate filaments subfamily represented in epithelial tissues.
Target Species Human, Mouse, Mammalia
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.