Learn More
Description
Specifications
Specifications
| Antigen | Cytokeratin 18 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SPM265 |
| Conjugate | PE |
| Dilution | Flow Cytometry |
| Gene Alias | Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 |
| Gene Symbols | KRT18 |
| Host Species | Mouse |
| Immunogen | PMC-42 human breast carcinoma cells |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
