Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 19 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP178278 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178278 0.1 mL
NBP178278SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP178278 Supplier Novus Biologicals Supplier No. NBP178278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Cytokeratin 19 Polyclonal specifically detects Cytokeratin 19 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 19
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Simple Western 1:3000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:400
Formulation PBS, 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. P08727
Gene Alias CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366
Gene Symbols KRT19
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Cytokeratin 19 protein (between residues 1-50) [UniProt P08727]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3880
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.