Learn More
Description
Specifications
Specifications
| Antigen | Cytokeratin 19 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.16 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/ml, Simple Western 1:3000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:400 |
| Formulation | PBS, 30% Glycerol with 0.05% Sodium Azide |
| Gene Accession No. | P08727 |
| Gene Alias | CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 |
| Gene Symbols | KRT19 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
