Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 19 Rabbit anti-Human, Mouse, Clone: KRT19/1959R, Novus Biologicals™
SDP

Catalog No. NB122860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122860 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122860 Supplier Novus Biologicals Supplier No. NBP30769320UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 19
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT19/1959R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, MGC15366
Host Species Rabbit
Immunogen Recombinant human full-length Cytokeratin 19 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3880
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.