Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 2 Rabbit anti-Human, Mouse, Rat, Clone: 9T3Q2, Novus Biologicals™
SDP

Catalog No. NB167370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167370 20 μg
NB167369 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167370 Supplier Novus Biologicals Supplier No. NBP31613920UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytokeratin 2 Monoclonal antibody specifically detects Cytokeratin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin 2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9T3Q2
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CK-2e, Cytokeratin-2e, epidermal ichthyosis bullosa of Siemens, Epithelial keratin-2e, K2e, keratin 2, keratin 2A (epidermal ichthyosis bullosa of Siemens), keratin, type II cytoskeletal 2 epidermal, Keratin-2 epidermis, keratin-2e, KRT2AMGC116967, KRT2EMGC116968, KRTE, Type-II keratin Kb2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 293-422 of human Cytokeratin 2 (P35908). YMIKVELQSKVDLLNQEIEFLKVLYDAEISQIHQSVTDTNVILSMDNSRNLDLDSIIAEVKAQYEEIAQRSKEEAEALYHSKYEELQVTVGRHGDSLKEIKIEISELNRVIQRLQGEIAHVKKQCKNVQD
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3849
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.