Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 20 Antibody (Ks20.8), Novus Biologicals™
SDP

Catalog No. NB120854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120854 0.5 mL
NB018095 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB120854 Supplier Novus Biologicals Supplier No. NB120854
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 20
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Ks20.8
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation Tissue culture supernatant with 0.05% Sodium Azide
Gene Accession No. P35900
Gene Alias CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT
Gene Symbols KRT20
Host Species Mouse
Immunogen Cytokeratins isolated from human micro-dissected villi of human duodenal mucosa.
Molecular Weight of Antigen 48.5 kDa
Purification Method Tissue culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 54474
Test Specificity This antibody reacts with cytokeratin polypeptide of 46 kDa in immunoblot analyses. Studies have shown that a marked difference exists in the expression of Cytokeratin 20 among various carcinomas.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.