Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 20 Antibody (Ks20.8), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $840.50
Specifications
Antigen | Cytokeratin 20 |
---|---|
Clone | Ks20.8 |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 20 | |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cytoskeleton Markers | |
P35900 | |
54474 | |
Cytokeratins isolated from human micro-dissected villi of human duodenal mucosa. | |
Primary | |
48.5 kDa |
Ks20.8 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT | |
KRT20 | |
IgG2a κ | |
Store at 4°C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title