Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 20 Antibody (OTI4A2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP271961A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271961A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271961A30 Supplier Novus Biologicals Supplier No. NBP271961AF350

Mouse Monoclonal Antibody

Cytokeratin 20 Monoclonal specifically detects Cytokeratin 20 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Cytokeratin 20
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4A2
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT
Gene Symbols KRT20
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-266 of human Cytokeratin 20 (NP_061883) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 54474.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.