Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 3/2p Antibody (2Q1040(AE5)), Novus Biologicals™
SDP

Catalog No. NBP23484405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23484405 0.05 mg
NBP23484425 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP23484405 Supplier Novus Biologicals Supplier No. NBP2348440.05MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 3/2p Monoclonal specifically detects Cytokeratin 3/2p in Human, Mouse, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Cytokeratin 3/2p
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2Q1040
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:50
Formulation PBS, pH 7.2, 0.5% BSA. with 0.09% sodium azide
Gene Alias 65 KDa Cytokeratin, CK3, CK-3, cytokeratin-3, Keratin, keratin 3, Keratin, Type II Cytoskeletal 3, Keratin-3, Type II Cytoskeletal 3, Type-II keratin Kb3
Gene Symbols KRT3
Host Species Mouse
Immunogen Rabbit corneal epithelial keratin
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 3850
Test Specificity Recognizes the 64kD polypeptide/keratin K3 (cytokeratin 3) of rabbit corneal epithelium and keratin K76 (cytokeratin K2p) of palate epithelium. As such represents an excellent marker for studying corneal epithelial-type differentiation. Also recognizes bovine snout and lip epithelium, as well as a rabbit corneal cell line.
Target Species Human, Mouse, Bovine, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.