Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 3/2p Antibody (2Q1040(AE5)), Novus Biologicals™

Mouse Monoclonal Antibody
$304.50 - $499.50
Specifications
Antigen | Cytokeratin 3/2p |
---|---|
Clone | 2Q1040 |
Dilution | Western Blot 1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:50 |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23484425
![]() |
Novus Biologicals
NBP2348440.025MG |
0.025 mg |
Each for $304.50
|
|
|||||
NBP23484405
![]() |
Novus Biologicals
NBP2348440.05MG |
0.05 mg |
Each for $499.50
|
|
|||||
Description
Cytokeratin 3/2p Monoclonal specifically detects Cytokeratin 3/2p in Human, Mouse, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Cytokeratin 3/2p | |
Western Blot 1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:50 | |
Monoclonal | |
Mouse | |
PBS, pH 7.2, 0.5% BSA. with 0.09% sodium azide | |
3850 | |
Rabbit corneal epithelial keratin | |
Primary | |
Recognizes the 64kD polypeptide/keratin K3 (cytokeratin 3) of rabbit corneal epithelium and keratin K76 (cytokeratin K2p) of palate epithelium. As such represents an excellent marker for studying corneal epithelial-type differentiation. Also recognizes bovine snout and lip epithelium, as well as a rabbit corneal cell line. |
2Q1040 | |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Human, Mouse, Bovine, Rabbit | |
65 KDa Cytokeratin, CK3, CK-3, cytokeratin-3, Keratin, keratin 3, Keratin, Type II Cytoskeletal 3, Keratin-3, Type II Cytoskeletal 3, Type-II keratin Kb3 | |
KRT3 | |
IgG1 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title