Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 3/2p Antibody (2Q1040(AE5)), PerCP, Novus Biologicals™
SDP

Catalog No. NB309986 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309986 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309986 Supplier Novus Biologicals Supplier No. NBP234844PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 3/2p Monoclonal antibody specifically detects Cytokeratin 3/2p in Human, Mouse, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen Cytokeratin 3/2p
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2Q1040(AE5)
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias 65 KDa Cytokeratin, CK3, CK-3, cytokeratin-3, Keratin, keratin 3, Keratin, Type II Cytoskeletal 3, Keratin-3, Type II Cytoskeletal 3, Type-II keratin Kb3
Host Species Mouse
Immunogen Rabbit corneal epithelial keratin
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3850
Target Species Human, Mouse, Bovine, Rabbit
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.