Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 3 Antibody (KRT3/2130), HRP, Novus Biologicals™
SDP

Catalog No. NB011105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011105 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011105 Supplier Novus Biologicals Supplier No. NBP279852H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT3/2130
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Alias CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3
Gene Symbols KRT3
Host Species Mouse
Immunogen Recombinant full-length human Cytokeratin 3 protein (Uniprot: P12035)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.