Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 3 Antibody (KRT3/2542) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119308 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119308 Supplier Novus Biologicals Supplier No. NBP308582
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT3/2542
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3
Host Species Mouse
Immunogen Tissue, cells or virus corresponding to Rabbit Cytokeratin 3/CK-3. Rabbit corneal epithelial keratin.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3850
Target Species Human, Bovine, Rabbit
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.