Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 4 Antibody (KRT4/2804), PerCP, Novus Biologicals™
SDP

Catalog No. NB385774 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385774 Supplier Novus Biologicals Supplier No. NBP308583PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 4 Monoclonal antibody specifically detects Cytokeratin 4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Cytokeratin 4
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone KRT4/2804
Conjugate PerCP
Formulation PBS
Gene Alias CK4, CK-4, CYK4cytokeratin-4, Cytokeratin-4, FLJ31692, K4cytokeratin 4, keratin 4, keratin, type II cytoskeletal 4, Keratin-4, Type-II keratin Kb4
Host Species Mouse
Immunogen Recombinant fragment (around aa 181-292) of human Cytokeratin 4 protein (exact sequence is proprietary) (Uniprot: P19013)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3851
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.