Learn More
Description
Specifications
Specifications
| Antigen | Cytokeratin5/14 |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | 16.4 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen 1-5 ug/ml |
| Formulation | PBS with 0.08% Sodium Azide |
| Gene Accession No. | P02533 |
| Gene Alias | 58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 |
| Gene Symbols | KRT5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
