Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 5/6/18 Antibody (LP34), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB386098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386098 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386098 Supplier Novus Biologicals Supplier No. NBP308545V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 5/6/18 Monoclonal antibody specifically detects Cytokeratin 5/6/18 in Human, Rat, Bovine samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin 5/6/18
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LP34
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias 58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5
Host Species Mouse
Immunogen Detergent-insoluble fraction of psoriatic human epidermis
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3852
Target Species Human, Rat, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.