Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 5/6 Antibody (A2A12), Novus Biologicals™
SDP

Catalog No. NB013473 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013473 Supplier Novus Biologicals Supplier No. NBP280401
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 5/6 Monoclonal specifically detects Cytokeratin 5/6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 5/6
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A2A12
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500Flow 1:50-1:100
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias 58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5
Gene Symbols KRT5
Host Species Mouse
Immunogen Synthetic peptide within Human Cytokeratin 5/6 aa 340-380. (SwissProt: P02538 Human; SwissProt: P04259 Human; SwissProt: P13647 Human)
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3852
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.