Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 5/8 Antibody (C-50), Novus Biologicals™
SDP

Catalog No. NBP24470401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24470401 0.1 mg
NBP24470400 0.02 mg
NBP24470402 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24470401 Supplier Novus Biologicals Supplier No. NBP2447040.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Cytokeratin 5/8 Monoclonal specifically detects Cytokeratin 5/8 in Human, Mouse, Rat, Porcine, Bovine, Canine, Hamster, Ferret, Sheep, Chicken (Negative), Xenopus (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen Cytokeratin 5/8
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone C-50
Concentration 0.2mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS (pH 7.4) and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P13647
Gene Alias cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5
Gene Symbols KRT5
Host Species Mouse
Immunogen Cytoskeletal preparation from HeLa cells
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3852
Test Specificity It reacts with keratin 5 (58kDa) and keratin 8 (52.5kDa). Simple epithelia express cytokeratin 8 in combination with 18. On the other hand, basal cells of stratified epithelia express cytokeratin 5 paired with 14. This antibody therefore, reacts with a wide range of epithelia and their carcinomas.
Target Species Human, Mouse, Rat, Pig, Canine, Ferret, Hamster, Primate, Sheep, Chicken (Negative), Xenopus (Negative)
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.