Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 5/8 Antibody (RCK102), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP197730A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP197730A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP197730A94 Supplier Novus Biologicals Supplier No. NBP197730AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 5/8 Monoclonal specifically detects Cytokeratin 5/8 in Human, Mouse, Rat, Porcine, Canine, Feline, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Cytokeratin 5/8
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RCK102
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5
Gene Symbols KRT5
Host Species Mouse
Immunogen Clone RCK102 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a mouse immunized with cytokeratins from a human lung cancer cell line (MR21).
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3852.0
Test Specificity RCK102 is reacting with cytokeratin 5 and cytokeratin 8. This monoclonal antibody recognizes virtually all epithelial tissues and carcinomas.
Target Species Human, Mouse, Rat, Pig, Canine, Feline, Hamster, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.