Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 5/8 Antibody (RCK102), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP197730J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197730J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197730J Supplier Novus Biologicals Supplier No. NBP197730APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 5/8 Monoclonal specifically detects Cytokeratin 5/8 in Human, Mouse, Rat, Porcine, Canine, Feline, Hamster, Rabbit samples. It is validated for Flow Cytometry.

Specifications

Antigen Cytokeratin 5/8
Applications Flow Cytometry
Classification Monoclonal
Clone RCK102
Conjugate APC
Dilution Flow Cytometry
Gene Alias cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5
Gene Symbols KRT5
Host Species Mouse
Immunogen Clone RCK102 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a mouse immunized with cytokeratins from a human lung cancer cell line (MR21).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3852
Test Specificity RCK102 is reacting with cytokeratin 5 and cytokeratin 8. This monoclonal antibody recognizes virtually all epithelial tissues and carcinomas.
Target Species Human, Mouse, Rat, Pig, Canine, Feline, Hamster, Rabbit
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.