Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 6 Antibody (KRT6B/2116), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB385572 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385572 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385572 Supplier Novus Biologicals Supplier No. NBP308612AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 6 Monoclonal antibody specifically detects Cytokeratin 6 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cytokeratin 6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT6B/2116
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6
Host Species Mouse
Immunogen Recombinant full-length human Cytokeratin 6 protein (Uniprot: P04259 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3853
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.