Learn More
Description
Specifications
Specifications
| Antigen | Cytokeratin 6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SPM269 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Alias | Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 |
| Gene Symbols | KRT6A |
| Host Species | Mouse |
| Immunogen | A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
