Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7/17 Antibody (C-46), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB50352A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50352A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50352A350 Supplier Novus Biologicals Supplier No. NB500352AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 7/17 Monoclonal specifically detects Cytokeratin 7/17 in Human, Porcine, Bovine, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen Cytokeratin 7/17
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C-46
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7
Gene Symbols KRT7
Host Species Mouse
Immunogen Cytoskeleton preparation of HeLa human cervix carcinoma cell line.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3855
Test Specificity The antibody C-46 reacts with Cytokeratin peptides 7 and 17 (54 and 46 kDa). Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues.
Target Species Human, Pig, Bovine, Mouse (Negative), Rabbit (Negative), Rat (Negative)
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.