Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7, 8 Antibody (LDS68), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP25046088 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25046088 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP25046088 Supplier Novus Biologicals Supplier No. NBP250460AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 7, 8 Monoclonal specifically detects Cytokeratin 7, 8 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Cytokeratin 7, 8
Applications Immunohistochemistry
Classification Monoclonal
Clone LDS68
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Cytoskeletons of the human urothelial RT4 cell line.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cellular Signaling
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.