Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7 Antibody (KRT7/903) - IHC-Prediluted, Novus Biologicals™
SDP

Catalog No. NBP248139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP248139 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP248139 Supplier Novus Biologicals Supplier No. NBP248139
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 7 Monoclonal specifically detects Cytokeratin 7 in Human, Ferret (Negative), Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 7
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT7/903
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7
Gene Symbols KRT7
Host Species Mouse
Immunogen Recombinant full-length human Cytokeratin 7 protein (Uniprot: P08729)
Molecular Weight of Antigen 54 kDa
Purification Method Protein A or G purified
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3855
Test Specificity Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-).
Target Species Human, Ferret (Negative), Mouse (Negative), Rabbit (Negative), Rat (Negative)
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.