Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7 Mouse anti-Human, Clone: OV-TL 12/30, Novus Biologicals™
SDP

Catalog No. NB153058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153058 0.2 mg
NB153057 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB153058 Supplier Novus Biologicals Supplier No. NBP3120130.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Cytokeratin 7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OV-TL 12/30
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7
Host Species Mouse
Immunogen This antibody was originally raised by immunizing BALB/c mice with ovarian carcinoma cell line OTN11.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.