Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 71 Antibody, Novus Biologicals™
SDP

Catalog No. NBP217040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217040 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217040 Supplier Novus Biologicals Supplier No. NBP217040
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytokeratin 71 Polyclonal antibody specifically detects Cytokeratin 71 in Human, Mouse, Acomys dimidiatus (spiny mouse), Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen Cytokeratin 71
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.91mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q3SY84
Gene Alias CK-71, cytokeratin-71, hK6irs, hK6irs1, K6IRS1MGC119390, K71, KB34, Keratin 6 irs, keratin 71, keratin, type II cytoskeletal 71, Keratin-71, KRT6IRS, KRT6IRS1MGC119391, Type II inner root sheath-specific keratin-K6irs1, Type-II keratin Kb34
Gene Symbols KRT71
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KRT71. The exact sequence is proprietary.
Molecular Weight of Antigen 57 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112802
Test Specificity Human. Expected to cross react based on sequence identity: Dog (91%), Bovine (91%).
Target Species Sheep, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.